ContactUsLinkComponent
Tissue Bond, Diagnostic Biosystems
Tissue Bond, Diagnostic Biosystems
# de catalogue CA99990-076
Fournisseur:  Diagnostic Biosystems
Numéro CAS:  
Tissue Bond, Diagnostic Biosystems
# de catalogue CA99990-076
Fournisseur:  Diagnostic Biosystems
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    TissueBond™ Reagent
  • Size:
    7 ml
  • Cat. no.:
    CA99990-076
  • Supplier No.:
    K 013
Spécifications
A propos de cet article
Proper tissue adhesion to the glass slides is critical in the immunostaining process. Improper adhesion of tissue sections can lead to poor quality staining or even loss of tissue sections during the process of immunostaining. White glue, albumen, chromegelatin and poly-L-lysine are used as tissue adhesives.
Glass treated with TissueBond™ can be used for both paraffin as well as frozen tissue sections. Since epitope recovery may require harsh treatment with proteolytic enzymes, microwave treatment or treatment with saponin etc., there is a need for a tissue adhesive which can irreversibly bind tissue sections to the glass without creating any background.
Customers Who Bought This Also Bought