Spécifications
- Description:TissueBond™ Reagent
- Size:7 ml
- Cat. no.:CA99990-076
- Supplier No.:K 013
Spécifications
A propos de cet article
Proper tissue adhesion to the glass slides is critical in the immunostaining process. Improper adhesion of tissue sections can lead to poor quality staining or even loss of tissue sections during the process of immunostaining. White glue, albumen, chromegelatin and poly-L-lysine are used as tissue adhesives.
Glass treated with TissueBond™ can be used for both paraffin as well as frozen tissue sections. Since epitope recovery may require harsh treatment with proteolytic enzymes, microwave treatment or treatment with saponin etc., there is a need for a tissue adhesive which can irreversibly bind tissue sections to the glass without creating any background.