Wire Gowning Benches, Eagle MHC
Fournisseur: Eagle Group
Certificats
A propos de cet article
Wire gowning bench wire shelving components that offer a more economical bench option
Ideal for less demanding environments or during construction as the room is brought into clean. Open wire construction facilitates 62% laminar flow.
Customers Who Bought This Also Bought