Mantle for Griffin-Type Beakers, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Heating mantles designed for use with Griffin-type beakers
Mantles are supplied complete with separable 121.9cm (4'), 2-wire cord and locking connector.
Certifications: CSA certified.
Customers Who Bought This Also Bought