Tweezers, 4" Wafer Handling, Excelta Corp®
Fournisseur: Excelta
Certificats
A propos de cet article
These tweezers are specifically designed handling 4" wafers.
- For scratch free holding of 4" wafers
- Cleanroom safe, anti-mag. Stainless steel
- Various configurations for various applications
The wafer tweezers for 4" wafers are made of stainless steel and are anti-magnetic. They come in a variety of configurations. The 'B' version features a 20° bend in the body to improve handling. The overall length is 4.75".
Customers Who Bought This Also Bought