Tweezers, Surface Mount, Excelta Corp®
Fournisseur: Excelta
Certificats
A propos de cet article
These tools are specifically designed to handle various electronic Surface Mount Devices.
- Specifically designed for specific applications
- Made of anti-magnetic stainless steel
- Fluted handles for better control
These S.M.D. paddle tip tweezers will firmly hold many types of SMD components. They are made of anti-magnetic Neverust® stainless steel and features serrated handles for better contol.
Customers Who Bought This Also Bought