Eyesaver® Eyewash Faucets, Speakman®
Fournisseur: Speakman
Certificats
A propos de cet article
Dedicated water ways allow the faucet and eyewash to work independently of each other.
- 'LIFT' activator for eyewash
- Aerated plastic spray outlets with flip-top dust caps
- Lead free brass construction
Model SEF-1850 is a single post laboratory faucet with an integrated ANSI compliant aerated eyewash.
Model SEF-1850-TW is a single post laboratory faucet with an integrated ANSI compliant aerated eyewash and tepid water system.
Model SEF-1850-4WH-TW is a single post laboratory faucet with an integrated ANSI compliant aerated eyewash and tepid water system. The faucet is equipped with 4" wrist blade handles.
Model SEF-1850-ST is a single post laboratory faucet with vacuum breaker and serrated tip. Includes an integrated ANSI compliant aerated eyewash.
Certifications: Meets ANSI Z358.1, ASME A112.18.1/CSA B125.1, NSF 61, CA AB 1953 and VT Act 193.
Customers Who Bought This Also Bought