SP Bel-Art Sterileware® Sampling Knife, Bel-Art Products, a part of SP
# de catalogue 56924-478
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Length:20.9 cm (8¼”)
- Cat. no.:56924-478
- Supplier No.:369650000
Spécifications
A propos de cet article
Ideal for cutting small samples from fibrous pieces, this sterile polystyrene plastic sampler knife has a blade that is serrated on one edge and smooth on the other.
The serrated edge is capable of cutting seafood, meat, poultry, vegetables, and other fibrous materials. Individually packaged, sterilized by gamma irradiation, lot stamped for accurate lot tracing, and shipped with a Certificate of Processing for Sterilization.
- Polystyrene plastic construction
- Sterile
- Blade is serrated on one edge and smooth on the other
- Sterilized via gamma irradiation
- Lot stamped for accurate lot tracing
The serrated edge is capable of cutting seafood, meat, poultry, vegetables, and other fibrous materials. Individually packaged, sterilized by gamma irradiation, lot stamped for accurate lot tracing, and shipped with a Certificate of Processing for Sterilization.
Customers Who Bought This Also Bought