Polypropylene Buckets, Contec®
Supplier: CONTEC, INC. MX
Certificates
About this item
Contec® bucket systems are available in a variety of size options for small and large critical environments. Replacement buckets are available for the polypropylene and stainless systems.
- Replacement buckets for polypropylene and stainless bucket systems
- For use with 3.9, 5, 6.5, and 7 gal. bucket systems
- Autoclavable
- Available in a variety of colors
These buckets are autoclavable and work with a variety of disinfectants and solvents. Wringer options are available based on bucket size.
Customers Who Bought This Also Bought