Spécifications
- Length:17.4 cm (6.8")
- Width:10 cm (3.9")
- Description:Mouse restrainer with scapular access, 3.2 cm
- Height:6.7 cm (2.6")
- Cat. no.:89136-652
- Weight Range:18-35 g
- Supplier No.:F464010000
Spécifications
A propos de cet article
This Mouse Restrainer with Dorsal Access takes care of the basics, so your hands are free for the important tasks.
- Easy-to-use design
- Two gate positions allow gentle and secure mouse immobilization
- Restrainer can be mounted to table or other surface
The easy-to-use entry funnel and tube with two gate positions allows quick, gentle and secure immobilization of the mouse. Draw mouse tail first, or lead head first, into the wide entry funnel and to the back of the tube, then place gate in choice of two slots. When in the restrainer, the dorsal skin of the mouse is accessible through a top circular opening so subcutaneous injections can be performed quickly, without risk of being bitten, and with less likelihood of sticking your own fingers. With tail first entry, the tail of the mouse is conveniently positioned outside the restrainer for tail bleeds and tattooing. Restrainer can be mounted to table or other surface. (Hardware not included). Disinfect with ethanol or common laboratory disinfectants, or autoclave to sterilize. Restrainer is made from durable, crystal clear TPX® and unlike acrylic restrainers will not craze or weaken.