SP Bel-Art Poxygrid Bag Holders, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
Sturdy wire holder keeps bags open while a separate hooked ring that rests over the top keeps them securely in place.
- Bright biohazard orange colored epoxy-coated steel wire
- Chemical- and corrosion-resistant
It will withstand a temperature range of –223 °C (–369 °F) to 121 °C (250 °F) and is autoclavable.
Customers Who Bought This Also Bought