SP Bel-Art Jerricans with Spigot, HDPE, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Space-saving, high-density polyethylene jerricans are great for dispensing without moving.
- HDPE construction
- Effectively dispense without moving
- Smooth-turning spigot
These models have a smooth-turning spigot that opens and closes with half a turn for easy, convenient dispensing from a stationary position.
Customers Who Bought This Also Bought