Anticon® Presaturated Wipes
# de catalogue 89092-960
Fournisseur: Contec
Small Business Enterprise
Spécifications
- W×L:22.9 x 22.9 cm (9 x 9")
- Description:Anticon® Gold Standard Weight Wipes, with 63% IPA/37% DI Water
- Cat. no.:89092-960
- Packaging:50/Bag, 16 Bags/Case
- Supplier No.:493650-239
Spécifications
A propos de cet article
Anticon® Presaturated Wipes are manufactured using ultra-clean Anticon® Gold and other Anticon® wipes in conjunction with semiconductor or USP grade solvents.
- Wipes 45800-088 and 45800-084 have a volume of 375 ml per bag
- Wipe 89092-960 has a volume of 60 ml per bag
These polyester knit cleanroom wipes are presaturated with the optimum amount of solution to deliver absolute cleaning power, not excess solution.
Anticon® presats are delivered with a patented low particle generation technology. Use Anticon® presats with the knowledge and security that there are no undo particle burdens being added to the critical environment.
Customers Who Bought This Also Bought