Keyed Quick Connect Plug, WaterSaver Faucet
Fournisseur: WaterSaver Faucet
Certificats
A propos de cet article
Plugs have an internal shut-off valve that closes when the valve is disengaged.
Internal valve components are stainless steel to assure compatibility with service to assure durable performance.
Certifications: CSA certified for natural gas.
Customers Who Bought This Also Bought