Hi-Lube Heavy-Duty Liquid Stirrer Lubricant, Ace Glass Incorporated
# de catalogue 89062-230
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Hi-Lube Heavy-Duty Liquid Stirrer Lubricant
- Cat. no.:89062-230
- Supplier No.:8119-07
Spécifications
A propos de cet article
Chlorofluorocarbon grease is designed for use at speeds up to 6000rpm with Ace Glass standard glass assemblies.
This lubricant has a high chemical inertness, and is unaffected by strong acids and alkalis. Soluble in most organic solvents. It is sitable for use with oxidizing gases. It also has high heat resistance, and is thermally stable up to 260°C (500°F). Non-flammable; does not carbonize on decomposition. Available in convenient one ounce (30mL) size.
Ordering information: For long stirring periods (over one hour), the use of a water-cooled type bearing is recommended. For information on other uses, please contact your Avantor representative.
Customers Who Bought This Also Bought