Stirrer Magnets, Octagonal, PTFE, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
These octagonal PTFE stirrer magnets feature a center pivot ring and are available in a wide variety of lengths and diameters
They are ideally suited for use in containers with bases that are slightly curved or uneven.
Customers Who Bought This Also Bought