Keyed Quick Connect Body, WaterSaver Faucet
Fournisseur: WaterSaver Faucet
Certificats
A propos de cet article
The quick connect keying system is based upon the inner and outer diameters of the sleeves on the body and plug
Quick connect bodies and plugs color coded to designate the service and to facilitate matching the correct body and plug.
Certifications: CSA certified for natural gas.
Customers Who Bought This Also Bought