To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Tubulin, Beta 3 Class Iii
- Clonality:Polyclonal
- Gene ID:10381
- Host:Chicken
- Isotype:IgY
- Reactivity:Human
- Antigen symbol:TUJ1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:50 μg
- Western blot:Yes
- Antigen synonyms:CDCBM|beta-4|CFEOM3A|FEOM3|CDCBM1|TUBB4|CFEOM3
- Storage buffer:PBS + sodium azide (0.02%, w/v)
- Molecular weight:50.43 kDa
- Storage temperature:4°C in the dark
- Concentration:100µg/ml
- Immunogen:Peptides
- Tested applications:ICC/IF, IHC, WB
- Purification:Antigen affinity purification
- Cat. no.:89353-938
- Supplier No.:GTX85469
Spécifications
A propos de cet article
Chicken polyclonal antibody to beta III Tubulin
Recommended starting dilutions: For IHC/IF: Use at 1:500 - 1:1000, for WB: Use at 1:1000 - 1:2500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: TUBB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat