To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Thyroid Hormone Receptor Interactor 230
- Clonality:Polyclonal
- Gene ID:9321
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:TRIP230
- Conjugation:Unconjugated
- ImmunoPrecipitation:Yes
- Size:50 μl
- Western blot:Yes
- Antigen synonyms:GMAP210|TRIP230|ACG1A|CEV14|TRIP-11|GMAP-210
- Storage buffer:Phosphate-buffered saline, pH 7.4, containing no preservatives
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Immunogen:Amino acids 1099-1372 of Trip230 expressed in E. coli.
- Tested applications:IP, WB
- Purification:Mouse Polyclonal Antiserum
- Cat. no.:89364-816
- Supplier No.:GTX70252
Spécifications
A propos de cet article
Mouse polyclonal antiserum to human TRIP230
For IP: Use at an assay dependent dilution. For WB: Use at a dilution of 1:5,000. Predicted molecular weight: 218 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: TRIP230
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human