To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Plasminogen Activator Inhibitor 1
- Clonality:Monoclonal
- Clone:3A120
- Gene ID:5054
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:PAI1
- Conjugation:Unconjugated
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:serpin family E member 1|PLANH1|PAI1|PAI|SERPINE1|PAI-1
- Storage buffer:50mM Sodium phosphate, 100mM sodium chloride,pH 7.4, containing no preservative
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:5.52 mg/ml
- Immunogen:Recombinant full length protein: human PAI
- Tested applications:WB
- Purification:Protein A purified
- Cat. no.:89363-366
- Supplier No.:GTX40924
Spécifications
A propos de cet article
Mouse monoclonal antibody [3A120] to PAI-1
WB: Use at an assay dependent dilution. Predicted molecular weight: 45 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: PAI1
Clonality: Monoclonal
Clone: 3A120
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human