Order Entry
ContactUsLinkComponent
Anti-TUBB3 Mouse Monoclonal Antibody [clone: TU-20]
Anti-TUBB3 Mouse Monoclonal Antibody [clone: TU-20]
# de catalogue 89362-648
Fournisseur:  Genetex
Mouse Monoclonal antibody to Neuron specific beta III Tubulin (tubulin, beta 3)
Anti-TUBB3 Mouse Monoclonal Antibody [clone: TU-20]
# de catalogue 89362-648
Fournisseur:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Antibody type:
    Primary
  • Antigen name:
    Tubulin Beta 3
  • Clonality:
    Monoclonal
  • Clone:
    TU-20
  • Gene ID:
    10381
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Baboon,
    Hamster,
    Carp,
    Pig,
    Mammals,
    Human,
    Bovine,
    Rat,
    Mouse
  • Antigen symbol:
    TUBB3
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Size:
    100 μg
  • Western blot:
    Yes
  • Antigen synonyms:
    Tubulin beta-3 chain|CDCBM|beta-4|CFEOM3A|FEOM3|CDCBM1|TUBB4|CFEOM3
  • Storage buffer:
    Phosphate buffered saline pH 7.4 containing 15mM sodium azide
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Immunogen:
    Synthetic peptide: ESESQGPK conjugated to malemide activated KLH, corresponding to amino acids 441-448 of Human Tubulin.
  • Tested applications:
    Fluorescence-Activated Cell Sorting
  • Purification:
    >95% by SDS-PAGE
  • Cat. no.:
    89362-648
  • Supplier No.:
    GTX27751

Spécifications

A propos de cet article

Mouse monoclonal antibody [TU-20] to beta III Tubulin

Flow Cytometry: Use at an assay dependant concentration
Western Blotting
Recommended dilution:1-2 µg/ml, 90 min
Positive control: Porcine brain lysate
Negative control: HPB-ALL human peripheral blood leukemia cell line
Sample preparation: Mix lysate with reducing Laemmli SDS-PAGE sample buffer.
Application note: Reducing conditions.
Immunohistochemistry (paraffin sections)
Recommended dilution: 10 µg/ml
Staining technique: Standard ABC technique (DAB+)
Pretreatment: 0.1% pepsin (trypsin) in 0.1 M HCl; incubation 30 min in RT; or Hightemperature citrate buffer antigen retrieval Positive tissue: neuronal tissue
Immunocytochemistry
Positive material: Neuro2a mouse neuroblastoma cell line
Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TUBB3
Clonality: Monoclonal
Clone: TU-20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Pig, Rat, Bb, Carp, Hamster, Mammals, Bovine

Customers Who Bought This Also Bought