To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) ligand 5
- Clonality:Polyclonal
- Gene ID:6352
- Host:Rabbit
- Isotype:IgG
- Neutralisation:Yes
- Reactivity:Human
- Antigen symbol:CCL5
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:50 μg
- Western blot:Yes
- Antigen synonyms:T-cell specific protein p288|normally T-expressed|C-C motif chemokine 5|t cell-specific protein P228|SIS-delta|small-inducible cytokine A5|small inducible cytokine subfamily A (Cys-Cys)|regulated upon activation|eoCP|and presumably secreted|beta-chemokine RANTES|T-cell-specific protein RANTES|member 5|eosinophil chemotactic cytokine|small inducible cytokine A5 (RANTES)
- Storage buffer:Phosphate buffered saline
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Immunogen:Recombinant human RANTES
- Tested applications:Functional Assay
- Purification:Purified IgG was prepared by antigen affinity chromatography.
- Cat. no.:89362-538
- Supplier No.:GTX27314
Spécifications
A propos de cet article
Rabbit polyclonal antibody to RANTES
Recommended Starting Dilutions:ELISA: 0.5 ug/ml - 2.0 ug/mlFunctional Assays: 5.0 ug/ml - 7.0 ug/mlWestern Blotting: 0.1 ug/ml - 0.2 ug/mlNot tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: CCL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: