To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:ATP Binding Cassette, Subfamily G, Member 2
- Clonality:Monoclonal
- Clone:BXP-21
- Gene ID:9429
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Antigen symbol:ABCG2
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:250 μl
- Western blot:Yes
- Format:Tissue culture supernatant
- Antigen synonyms:EST157481|MRX|MXR-1|GOUT1|sub-family G (WHITE)|MXR1|BCRP1|BCRP|CDw338|MXR|member 2 (Junior blood group)|ABC15|CD338|BMDP|ATP binding cassette subfamily G member 2 (Junior blood group)|ATP-binding cassette|ABCP|UAQTL1
- Amino acid number:271 to 396
- Storage buffer:Preservative: 0.1% Sodium Azide; Constituents: 1% BSA
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C.
- Immunogen:Fusion protein composed of E. Coli maltose binding protein and BCRP peptide [126 amino acids, 271-396 AA of BCRP, GeneBank accession #AF098951].
- Tested applications:ICC/IF, IHC-P, IHC-Fr, WB
- Purification:Tissue culture supernatant
- Cat. no.:89352-006
- Supplier No.:GTX23380
Spécifications
A propos de cet article
Mouse Monoclonal antibody [BXP-21] to ABCG2
ICC: Use at a dilution of 1/20 - 1/50. (Acetone fixed cells) IHC-P: Use at a dilution of 1/20 - 1/40. (Pre treatment with citrate buffer pH 6.0 is required)IHC-Fr: Use at a dilution of 1/20 - 1/40. WB: Use at an assay dependent dilution. Not tested in other applications.
Type: Primary
Antigen: ABCG2
Clonality: Monoclonal
Clone: BXP-21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human