To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Fibronectin
- Clonality:Polyclonal
- Gene ID:2335
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:FN1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:Cold insoluble globulin|ED-B|FNZ|MSF|CIG|GFND2|LETS|fibronectin 1|GFND|FN|FINC
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 and 0.01% (w/v) Sodium Azide
- Storage temperature:For extended storage mix with an equal volume of glycerol, aliquot contents and freeze at –20 °C or below.
- Concentration:1.0 mg/ml
- Immunogen:Fibronectin was purified from Human plasma by binding to a denatured gelatin column followed by elution with high concentrations of arginine.? The eluted material was further purified by gel filtration.? Immunization occurred after single-band purity was
- Tested applications:ELISA, IHC-P, IP, WB
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89360-822
- Supplier No.:GTX20299
Spécifications
A propos de cet article
Fibronectin is a glycoprotein present in a soluble dimeric form in plasma, and in a dimeric or multimeric form at the cell surface and in extracellular matrix. Fibronectin is involved in cell adhesion and migration processes including embryogenesis, wound healing, blood coagulation, host defense, and metastasis. They occur as dimers of two 200 kDa subunits. They have binding domains for bacterial proteins, collagens, heparin-like molecules and fibrin. Cellular fibronectin is widely distributed in the stroma of malignant tumours.
For ELISA: Use at a dilution of 1:4,000-1:8,000. For IHC-P: Use at a dilution of 1:50-1:200. For IP: Use at an assay dependent dilution. For WB: Use at a dilution of 1:4,000-1:8,000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: FN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human