To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Tubulin beta
- Clonality:Polyclonal
- Gene ID:203068
- Host:Rabbit
- Isotype:IgG
- Reactivity:Xenopus,Chimpanzee,Human,Rat,Mouse
- Antigen symbol:TUBB
- Conjugation:Unconjugated
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:tubulin beta polypeptide|tubulin beta-5 chain|M40|beta Ib tubulin|tubulin|beta 5-tubulin|tubulin beta chain|tubulin beta-1 chain|CDCBM6|beta polypeptide|beta-4 tubulin|OK/SW-cl.56|TUBB1|TUBB5
- Storage buffer:Preservative: 0.1% Sodium Azide. Constituents: 0.2% Gelatin, PBS
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:Synthetic peptide (Human) (N terminal).
- Tested applications:WB
- Purification:Immunogen affinity purified
- Cat. no.:89351-462
- Supplier No.:GTX18587
Spécifications
A propos de cet article
Tubulin is the major building block of microtubules. This intracellular cylindrical filamentous structure is present in almost all eukaryotic cells. Microtubules function as structural and mobile elements in mitosis, intracellular transport, flagellar movement, and in the cytoskeleton.
WB: 1/400. Predicted molecular weight: 49 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: TUBB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chimpanzee