Order Entry
ContactUsLinkComponent
Anti-SYP Mouse Monoclonal Antibody [clone: 4E+206]
Anti-SYP Mouse Monoclonal Antibody [clone: 4E+206]
# de catalogue 89360-366
Fournisseur:  Genetex
Anti-SYP Mouse Monoclonal Antibody [clone: 4E+206]
# de catalogue 89360-366
Fournisseur:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Antibody type:
    Primary
  • Antigen name:
    Synaptophysin
  • Clonality:
    Monoclonal
  • Clone:
    4E206
  • Gene ID:
    6855
  • Host:
    Mouse
  • Isotype:
    IgM
  • Reactivity:
    Bovine
  • Antigen symbol:
    SYP
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Size:
    50 μl
  • Western blot:
    Yes
  • Antigen synonyms:
    MRXSYP|major synaptic vesicle protein P38|MRX96
  • Storage buffer:
    Preservative: None. Constituents: 50% Glycerol, PBS, 0.1mM PMSF. pH 7.2
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Immunogen:
    Synaptic vesicle-containing fractions immunoprecipitated from human brain homogenates using anti-human Synaptophysin monoclonal antibodies.
  • Tested applications:
    IHC, IP, WB
  • Purification:
    Purified IgM
  • Cat. no.:
    89360-366
  • Supplier No.:
    GTX18008

Spécifications

A propos de cet article

Synaptophysin is a 38kDa glycoprotein present in the membrane of neuronal presynaptic vesicles in brain, spinal cord, retina, vesicles of adrenal medulla, neuromuscular junctions, and endocrine cells. It is also expressed by neuroendocrine cells throughout the body, both normal and neoplastic. Synaptophysin is a useful marker for the identification of normal neuroendocrine cells and neuroendocrine neoplasms. This antibody reacts with neuroendocrine cells of human adrenal medulla, carotid body, skin, pituitary gland, thyroid, lung, pancreas, and gastointestinal mucosa. It also reacts with a wide spectrum of neuroendocrine neoplasms of neural type including neuroblastomas, ganglioneuroblastomas, ganglioneuromas, pheochromocytomas, chromaffin, and non-chromaffin paragangliomas.

IHC: Use at an assay dependent dilution. IP: 1/100. WB: 1/2000. Detects a band of approximately 38 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SYP
Clonality: Monoclonal
Clone: 4E+206
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Bovine

Customers Who Bought This Also Bought