Order Entry
ContactUsLinkComponent
Anti-M13 Mouse Monoclonal Antibody
Anti-M13 Mouse Monoclonal Antibody
# de catalogue 89360-274
Fournisseur:  Genetex
Anti-M13 Mouse Monoclonal Antibody
# de catalogue 89360-274
Fournisseur:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Antibody type:
    Primary
  • Antigen name:
    M13 bacteriophage
  • Clonality:
    Monoclonal
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Antigen symbol:
    M13
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • Western blot:
    Yes
  • Storage buffer:
    Phosphate-buffered saline containing 0.08% sodium azide
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Immunogen:
    Full length M13 phage coat protein; Exact epitope not determined.
  • Tested applications:
    Fluorescence-Activated Cell Sorting
  • Purification:
    Protein A purified
  • Cat. no.:
    89360-274
  • Supplier No.:
    GTX17269

Spécifications

A propos de cet article

M13 antibody [E1] (GTX17269) has numerous uses in screening phage display libraries, especially the screening of single chain antibody libraries.

Type: Primary
Antigen: M13
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:

Customers Who Bought This Also Bought