To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Erb-b2 receptor tyrosine kinase 4
- Clonality:Polyclonal
- Gene ID:2066
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:ERBB4
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:500 μl
- Western blot:Yes
- Antigen synonyms:ERBB4|Verb a e|Tyrosine kinase type cell surface receptor HER4|p180erbB4|Avian erythroblastic leukemia viral (verbb2) oncogene homolog 4|C-erbB-4
- Storage buffer:Phosphate-buffered saline, pH 7.4 containing 1% BSA and 0.09% sodium azide
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:Synthetic peptide (unfortunately, the amino acid sequence is considered to be commercially sensitive) (Human) (C terminal).
- Tested applications:IHC-P, IP, WB
- Purification:Immunogen affinity purified
- Cat. no.:89350-958
- Supplier No.:GTX15137
Spécifications
A propos de cet article
ErbB 4 is the fourth member of class I receptor kinase family. EbbB 4 is most predominantly expressed in several breast carcinoma cell lines, and in normal skeletal muscle, heart, pituitary, brain, and cerebellum. Breast tumor cell lines T47-D, MDA-MB-453, BT-474 and H3396 are found to have the highest levels of mRNA, and intermediate levels are seen in MCF-7, MDAMB-330 and MDA-MB-361. Expression of ErbB 4 is low or absent in some breast tumor cell lines such as MDA-MB-231, MDA-MB-157, MDA-MB-468, and SKBR-3.
IHC-P: Use at a dilution of 1/50 for 10 min at RT. Boil tissue sections in 10mM citrate buffer, pH 6.0 for 10 min followed by cooling at RT for 20 min. IP: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 146.8 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: ERBB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human