Order Entry
ContactUsLinkComponent
Anti-LHX3 Rabbit Polyclonal Antibody
Anti-LHX3 Rabbit Polyclonal Antibody
# de catalogue 89360-036
Fournisseur:  Genetex
Anti-LHX3 Rabbit Polyclonal Antibody
# de catalogue 89360-036
Fournisseur:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Antibody type:
    Primary
  • Antigen name:
    LIM Homeobox 3
  • Clonality:
    Polyclonal
  • Gene ID:
    16871
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Fish
  • Antigen symbol:
    LHX3
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Size:
    50 μg
  • Western blot:
    Yes
  • Antigen synonyms:
    DKFZp762A2013|CPHD3|LIM3|M2-LHX3
  • Storage buffer:
    Preservative: 0.1% Sodium Azide; Constituents: PBS. pH 7.2
  • Storage temperature:
    Add glycerol to a final volume of 50%, aliquot and store at –20 °C. Avoid repeated freeze / thaw cycles.
  • Concentration:
    0.5 mg/ml
  • Immunogen:
    C-terminal portion of mouse LHX3 protein.
  • Tested applications:
    ICC/IF, IHC-P, IHC-Fr, IP, WB
  • Purification:
    Protein A purified
  • Cat. no.:
    89360-036
  • Supplier No.:
    GTX14555

Spécifications

A propos de cet article

The pituitary gland releases hormones that regulate growth, lactation, homeostasis, the stress response, reproductive development and fitness as well as water metabolism. LHX3 is one of several transcription factors that have been implicated in pituitary development. This particular transcription factor is critical for motor neuron specification and the development of the anterior and intermediate lobes of the pituitary gland in mammals. The sequence of the human LHX3 gene has been identified and characterized by IU researchers.

ICC: Use at a dilution of 1/100 - 1/250. IF: Use at an assay dependent dilution. Use a secondary antibody conjugated to biotin and then streptavidin-conjugated to fluorochrome. IHC-P: Use at a dilution of 1/125 - 1/500. IHC-Fr: 1:1500-1:3000. For whole mounts recommended fixative is MEMFA using a dilution of 1/2000 - 1/4000. IP: Use at a dilution of 1/100. WB: Use at a dilution of 1/1250 - 1/2500. Predicted molecular weight: 44 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: LHX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Fish

Customers Who Bought This Also Bought