To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Toll-like Receptor 7
- Clonality:Polyclonal
- Gene ID:51284
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:TLR7
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:50 μg
- Western blot:Yes
- Antigen synonyms:TLR7|UNQ248|Toll-like receptor 7|Anti-TLR7|PRO285
- Storage buffer:PBS and 0.05% sodium azide was added as a preservative.
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Synthetic peptide: LSHNQLTTVPERLSNCSRSLKNL conjugated to KLH, corresponding to amino acids 706-728 of Human TLR7.
- Tested applications:Fluorescence-Activated Cell Sorting
- Purification:Protein G purified
- Cat. no.:89359-836
- Supplier No.:GTX13730
Spécifications
A propos de cet article
The Toll like receptor (TLR) family in mammal comprises a family of transmembrane proteins characterized by multiple copies of leucine rich repeats in the extracellular domain and IL 1 receptor motif in the cytoplasmic domain. Like its counterparts in Drosophila, TLRs signal through adaptor molecules and could constitute an important and unrecognized component of innate immunity in humans. The TRL family is a phylogenetically conserved mediator of innate immunity that is essential for microbial recognition. TLRs characterized so far activate the MyD88/interleukin 1 receptor-associated kinase (IRAK) signaling pathway. Ten human homologs of TLRs (TLR1 10) have been described. Stimulation of the NFkB signaling pathway by TLR7 suggests that it plays a role in immune response.
We recommend the following starting dilutions:
For WB: Use at a concentration of 1-3 ug/ml.
For ICC/IF: Use at a dilution of 1:10-1:2000.
For FACS: Use at a concentration of 2-5 ug/ 1x10^6 cells.
For IHC-Fr: Use at a dilution of 1:10-1:2000.
For IHC-P: Use at a assay dependent dilution.
Optimal dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: TLR7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human