To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Psoriasin
- Clonality:Monoclonal
- Clone:47C1068
- Gene ID:6278
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:PSOR1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:S100A7c|psoriasin 1|S100 calcium-binding protein A7 (psoriasin 1)|protein S100-A7|PSOR1
- Storage buffer:Preservative: 0.02% Sodium Azide; Constituents: 0.2% Gelatin, PBS
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Recombinant full length protein (Human).
- Tested applications:IHC, WB
- Purification:Protein G purified
- Cat. no.:89359-812
- Supplier No.:GTX13680
Spécifications
A propos de cet article
Psoriasin/HID-5 was originally identified as an up-regulated protein in psoriatic keratinocytes. Subsequently, this protein was found to have elevated expression in abnormally differentiating primary keratinocytes and in various carcinomas, including ductal carcinoma in situ. Psoriasin/HID-5 may play a role in the establishment or maintenance of these pathologies, and may serve as a diagnostic marker of particular tumour types or abnormally proliferative epithelia.
IHC: Use at an assay dependent dilution. WB: Use at a concentration of 1 ?g/ml. Detects a band of approximately 11 kDa (predicted molecular weight: 12.1 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: PSOR1
Clonality: Monoclonal
Clone: 47C1068
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human