Order Entry
ContactUsLinkComponent
Anti-TAP1 Rabbit Polyclonal Antibody
Anti-TAP1 Rabbit Polyclonal Antibody
# de catalogue 89359-714
Fournisseur:  Genetex
Anti-TAP1 Rabbit Polyclonal Antibody
# de catalogue 89359-714
Fournisseur:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Antibody type:
    Primary
  • Antigen name:
    Transporter 1, ATP Binding Cassette Subfamily B member
  • Clonality:
    Polyclonal
  • Gene ID:
    6890
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Antigen symbol:
    TAP1
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Size:
    50 μl
  • Western blot:
    Yes
  • Antigen synonyms:
    ABC17|RING4|PSF1|FLJ41500|ABCB2|TAP1*0102N|TAP1N|D6S114E|DAAP-57C1.5|APT1|FLJ26666
  • Storage buffer:
    Preservative: NoneConstituents: Whole Serum
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Immunogen:
    Synthetic peptide: AKLVQRQMLGLQPA, corresponding to amino acids 735-748 of Human TAP1.
  • Tested applications:
    ICC/IF, IP, WB
  • Purification:
    Whole antiserum
  • Cat. no.:
    89359-714
  • Supplier No.:
    GTX13516

Spécifications

A propos de cet article

The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The protein encoded by this gene is involved in the pumping of degraded cytosolic peptides across the endoplasmic reticulum into the membrane-bound compartment where class I molecules assemble. Mutations in this gene may be associated with ankylosing spondylitis, insulin-dependent diabetes mellitus, and celiac disease.

ICC: Use at an assay dependent dilution. IP: Use at a dilution of 1/100. WB: Use at a dilution of 1/4000. Detects a band of approximately 70 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Customers Who Bought This Also Bought