To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Bassoon
- Clonality:Monoclonal
- Clone:SAP7F407
- Gene ID:29138
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Mouse
- Antigen symbol:Bsn
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:50 μg
- Western blot:Yes
- Storage buffer:Preservative: None. Constituents: 50% Glycerol, PBS, 0.1mM PMSF
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant rat bassoon fragment (residues 738-1035) expressed asa fusion protein in E.coli.
- Tested applications:ICC/IF, IHC, IP, WB
- Purification:Protein G purified
- Cat. no.:89359-614
- Supplier No.:GTX13249
Spécifications
A propos de cet article
Bassoon is a 420 kDa protein that is a localized at the presynaptic nerve terminals and is believed to play a role in the structural and functional organization of the synaptic vesicle cycle. Bassoon does not belong to any known protein families. It has been found in rat and mouse with sequence identity of the two proteins at 96%. The human BASSOON gene has recently been cloned and localized. Bassoon is predicted to contain two double-zinc fingers, three coiled-coil regions, and two polyglutamine domains. The polyglutamine domains in the C-terminus are of interest, since it is known that for some human proteins, such as Huntingtin, abnormal amplification of this region can cause late-onset neurodegeneration. Bassoon is concentrated at sites opposite to postsynaptic densities in synaptic terminals and in cultured neurons, it is found to colocalize with GABA (A) and glutamate (GluR1) receptors.Another presynaptic protein, Piccolo was found to colocalize with Bassoon in cultured hippocampal neurons. These observations suggested that they serve specific functions at synaptic junctions and may be involved in organization of the cytoskeleton at the site of neurotransmitter release.
ICC: Use at a concentration of 0.5 - 5 ?g/ml. IHC: Use at a concentration of 2 ?g/ml. IP: Use at an assay dependent dilution. WB: Use at a concentration of 1 ?g/ml. Detects a band of approximately 400 kDa (predicted molecular weight: 472.6 kDa). Additional bands between 97 and 400 kDa are also identified and are reported to be proteolytic degradation products of Bassoon. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Bsn
Clonality: Monoclonal
Clone: SAP7F407
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Rat