Order Entry
ContactUsLinkComponent
Anti-PRMT5 Mouse Monoclonal Antibody [clone: PRMT5-21]
Anti-PRMT5 Mouse Monoclonal Antibody [clone: PRMT5-21]
# de catalogue 89359-248
Fournisseur:  Genetex
Anti-PRMT5 Mouse Monoclonal Antibody [clone: PRMT5-21]
# de catalogue 89359-248
Fournisseur:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein Arginine Methyltransferase 5
  • Clonality:
    Monoclonal
  • Clone:
    PRMT5-21
  • Gene ID:
    10419
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Bovine
  • Antigen symbol:
    PRMT5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Size:
    50 μl
  • Western blot:
    Yes
  • Antigen synonyms:
    SKB1|HRMT1L5|IBP72|SKB1Hs|JBP1
  • Storage buffer:
    Preservative: 15mM Sodium Azide; Constituents: 0.01M PBS. pH 7.4
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    2 mg/ml
  • Immunogen:
    Antibody was produced by the fusion of mouse myeloma cells and splenocytes from mice immunized with PRMT5 containing the C-terminal region of the Human PRMT5 (amino acids 315-637).
  • Tested applications:
    ELISA, WB
  • Purification:
    Protein A purified
  • Cat. no.:
    89359-248

Spécifications

A propos de cet article

Arginine methylation is an irreversible post translational modification which has only recently been linked to protein activity. At least three types of PRMT enzymes have been identified in mammalian cells. These enzymes have been shown to have essential regulatory functions by methylation of key proteins in several fundamental areas. These protein include nuclear proteins (Histone 2A, 3, 4), IL enhancer binding factor, nuclear factors (NF45, 90, ILF3, Nucleolin, STAT1, Poly(A) binding protein II), cell cycle proteins (phosphoprotein phosphatase 2A), signal transduction proteins (FGF2, Fibrillarin, FN, INFAR1, Jak, MBP, Src-adaptor Sam68), apoptosis proteins (FADD, ICE-like protease), and viral proteins (Hepatitis C NS3 RNA Helicase, HIV TAR). The mammalian PRMT family currently consists of 5 members that share two large domains of homology. Outside of these domains, epitopes were identified and antibodies against all five PRMT members have been developed. These antibodies can be utilized to explore arginine methylation and its regulatory functions.

ELISA: Use at an assay dependent dilution. WB: Use at a concentration of 2 ?g/ml (determined by using a whole cell extract of human melanoma cell line G361). Predicted molecular weight: 70 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PRMT5
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine

Customers Who Bought This Also Bought