To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Splicing Factor Proline/glutamine-rich
- Clonality:Monoclonal
- Clone:B92
- Gene ID:6421
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:SFPQ
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:50 μl
- Western blot:Yes
- Antigen synonyms:hPOMp100|splicing factor proline/glutamine rich (polypyrimidine tract-bindin|PTB-associated splicing factor|polypyrimidine tract-binding protein-associated splicing factor|polypyrimidine tract binding protein associated|PTB-associated-splicing factor|splicing factor proline/glutamine rich (polypyrimidine tract binding protein associated)|DNA-binding p52/p100 complex|polypyrimidine tract-binding protein-associated-splicing factor|PSF|100 kDa subunit|POMP100|100 kDa DNA-pairing protein
- Storage buffer:Phosphate-buffered saline, pH 7.4, containing 15 mM sodium azide
- Storage temperature:Upon Receipt - Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:2.1 mg/ml
- Immunogen:Tissue / cell preparation (lysate of mouse bone marrow-derived stromal cell line.).
- Tested applications:Affinity Purification, Radio Immunoassay
- Purification:Protein A purified
- Cat. no.:89358-864
- Supplier No.:GTX11825
Spécifications
A propos de cet article
The RNAs that direct protein synthesis in animals and plant cells are synthesized in the nucleus as large precursors (pre-mRNAs). The protein coding sequences in pre-mRNA molecules are arranged in discontinuous segments - exons interspersed with noncoding sequences - introns. In a process termed splicing, these introns are efficiently removed before the pre-mRNA is transported from the nucleus to the cytoplasm, where it is translated into protein. Studies have shown that nuclear pre-mRNA splicing takes place in a multi-component structure termed a spliceosome. The polypyrimidine tract-binding (PTB) protein-associated splicing factor (PSF), which plays an essential role in mammalian spliceosomes, is a ubiquitous nuclear matrix protein. A complex between PTB and PSF is necessary for pre-mRNA splicing. PSF contains two consensus RNA-binding domains and an unusual amino terminus rich in proline and glutamine residues. The RNA-binding properties of PSF are apparently identical to those of PTB. Both proteins, together and independently, bind the polypyrimidine tract of mammalian introns. However, the nuclear localization of PSF and PTB and their distribution in subnuclear fractions differ markedly: isolated nuclear matrices contain a bulk of PSF, but only minor amounts of PTB. In confocal microscopy both proteins appear in speckles, the majority of which do not co-localize. These PTB/PSF complexes, as well as the observed PSF-PTB interaction, may reflect the presence of PTB and PSF in spliceosomal complexes during RNA processing, although other data point to different cellular distribution and nuclear matrix association of the majority of PSF and PTB. The cleavage of PSF during lysis of immature myeloid cells is accompanied by digestion of the PTB splicing regulator but not other proteins tested. In contrast, during apoptosis PTB is degraded while PSF remains intact. Proteolytic degradation of PSF specifically occurs in intact myeloid cells and this process is enhanced upon immature myeloid cell lysis; PSF is completely cleaved to a 47 kDa proteolytic cleavage product (p47), due to potent proteolytic activity found in these cells but rare in other cells and tissues. Furthermore, p47 is abundant in intact normal and tumor myeloid cells while in other cell types it is undetectable. The bone marrow 47 kDa protein is a fragment constituting the N-terminal, protease-resistant half of the splicing factor PSF. PSF is highly basic and migrates anomalously on SDS gels. The 47 kDa protein of mouse cells of immature myeloid origin (bone marrow and acute myeloid leukemia) exhibits a gel migration pattern corresponding to a 49 kDa molecule. In other cell types such as lymphoid cells and in peripheral blood cells, PSF appears as approx. 100 kDa or 75 kDa molecules. The sequence of a fragment of mouse PSF was found to be remarkably similar to that of human PSF ( > 98% homology). Also, the sequences of PSF and the human (h) 100 kDa DNA-pairing protein (hPOMp100) reveals identity. Homologous pairing is a fundamental biological reaction implicated in various cellular processes such as DNA recombination and repair, chromosome pairing, sister chromatid cohesion and chromosome condensation, gene inactivation and initiation of replication. The base pairing is also involved in spliceosome assembly resulting in formation of a dynamic Holliday-like structure within which splicing occurs. Indeed, PSF/hPOMp100 bind both singlestranded (ss) and double-stranded (ds) DNA and facilitates the renaturation of complementary ssDNA molecules. Importantly, PSF/hPOMp100 promotes the formation of D-loops in superhelical duplex DNA. PSF/hPOMp100 also serves as an efficient substrate for protein kinase C (PKC) in vitro. PKC phosphorylation of PSF/hPOMp100 stimulates its DNA binding and D-loop formation activity suggesting a possible regulatory mechanism. PSF has been demonstrated to interact with a variety of cellular targets including the human pro-oncoproteins EWS, hPOMp75/TLS and calmodulin, the RNA/DNAbinding nuclear protein p54nrb/NonO (the homolog of PSF) and DNA topoisomerase. A direct interaction has been observed, between PSF and topoisomerase I which has been implied in DNA recombination, DNA repair, and chromosome formation and may act as a transcription factor and a protein kinase. PSF is also expressed by differentiating neurons in developing mouse brain. Both the expression of PSF mRNA in cortex and cerebellum and PSF immunoreactivity in all brain areas has been found to be high during embryonic and early postnatal life. In adult tissue, only various neuronal populations in the hippocampus and olfactory bulb express PSF. PSF is expressed by differentiating neurons but not by astrocytic cells including radial glia; however oligodendrocytes differentiating in vitro were found to express it. The restricted expression of PSF suggests that it is involved in the control of neuronal-specific splicing events occurring at particular stages of neuronal differentiation and maturation. Monoclonal antibodies reacting specifically with PSF are useful tools for the molecular identification and characterization of the functional activity of PSF.
For AP: Use at an assay dependent dilution (see Shav-Tal et al reference). For ICC: Use at an assay dependent dilution (fix cells 2 min in 3% paraformaldehyde with 0.5% Triton X-100, and then 20 min in 3% paraformaldehyde) (see Lee et al reference). For IHC-Fr: Use at an assay dependent dilution (4% paraformaldehyde-0.5% Triton X-100 treated tissue). For IP: Use at an assay dependent dilution (see Lee et al reference). For RIA: Use at an assay dependent dilution (see Lee et al reference). WB: WB: Use at a concentration of 2 - 4 ?g/ml. Detects a band of approximately 47, 49, 100, 68, 120, 100, 80 and 68 kDa (predicted molecular weight: 76 kDa). (See Lee et al reference, Shav-Tal et al reference). Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: SFPQ
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human