To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Tubulin, Gamma 1
- Clonality:Polyclonal
- Gene ID:7283
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:TUBG1
- Conjugation:Cy3®
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 μl
- Antigen synonyms:TUBGCP1|CDCBM4|TUBG|GCP-1
- Storage buffer:Preservative: 15mM Sodium Azide; Constituents: 1% BSA, 0.01M PBS. pH 7.4
- Concentration:1.3mg/ml
- Immunogen:Synthetic peptide: EEFATEGTDRKDVFFY-K conjugated to KLH, corresponding to amino acids 38-53 of Human gamma Tubulin.
- Tested applications:ICC/IF
- Purification:Immunogen affinity purified
- Cat. no.:89357-366
- Supplier No.:GTX11319
Spécifications
A propos de cet article
Rabbit polyclonal to gamma Tubulin - Centrosome Marker ( Cy3 )
IF: Use at a dilution of 1/200. This concentration was determined by staining of methanol/acetonefixed chicken fibroblast cells. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: TUBG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human