Order Entry
ContactUsLinkComponent
Anti-SPTA1 Mouse Monoclonal Antibody [clone: SB-SP1]
Anti-SPTA1 Mouse Monoclonal Antibody [clone: SB-SP1]
# de catalogue 89356-866
Fournisseur:  Genetex
Anti-SPTA1 Mouse Monoclonal Antibody [clone: SB-SP1]
# de catalogue 89356-866
Fournisseur:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Antibody type:
    Primary
  • Antigen name:
    Spectrin
  • Clonality:
    Monoclonal
  • Clone:
    SB-SP1
  • Gene ID:
    6708
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Antigen symbol:
    SPTA1
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Size:
    100 μl
  • Western blot:
    Yes
  • Format:
    Ascites
  • Antigen synonyms:
    SPH3|HS3|SPTA|EL2|HPP
  • Storage buffer:
    Preservative: 0.1% Sodium Azide; Constituents: Raw Ascites
  • Storage temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Immunogen:
    Full length native Human erythroid spectrin was used as the immunogen (purified).
  • Tested applications:
    ICC/IF, WB
  • Purification:
    Ascites
  • Cat. no.:
    89356-866
  • Supplier No.:
    GTX11184

Spécifications

A propos de cet article

Mouse monoclonal [SB - SP1] to Spectrin

IF: Use at an assay dependent dilution. By immunofluorescence the antibody will stain human spectrin in erythrocyte smears. WB: Use at a dilution of 1/500 (determined using an extract of human erythrocyte ghosts or purified human erythrocyte spectrin and immunoperoxidase labeling). Expected bands at 220 kDa (beta) and 240 kDa (alpha) chains of human erythrocyte spectrin. The antibody stains the beta subunit stronger than the alpha subunit at the recommended titer. In addition, a number of polypeptides in the region of 170, 150, 85, and 45 kDa will react with the antibody. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SPTA1
Clonality: Monoclonal
Clone: SB-SP1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Customers Who Bought This Also Bought