To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Alpha 1 Fetoprotein Receptor
- Clonality:Monoclonal
- Clone:AFP-01
- Gene ID:174
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:AFP
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoPrecipitation:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:FETA|Alpha-1-Fetoprotein|HPAFP
- Storage buffer:Phosphate buffered saline pH 7.4 containing 15mM sodium azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Full length native protein (purified) (Human)
- Tested applications:ELISA, IP, WB
- Purification:>95% by SDS-PAGE
- Cat. no.:89361-414
Spécifications
A propos de cet article
Mouse monoclonal [AFP - 01] to alpha 1 Fetoprotein
For ELISA: Use at an assay dependant dilution. The antibody AFP-01 is suitable for sandwich-ELISA with the antibody AFP-11. For IP: Use at an assay dependant dilution. Interaction of the antibody with AFP is dependent on the presence of calcium ions (strongly inhibited by chelating agents). Such characteristics of the antibody can be exploited for immunoaffinity purification of APF under mild elution conditions. For WB: Use at an assay dependant dilution. Detects a band of approximately 70 kDa. Use under non-reducing conditions. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: AFP-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human