Septa, Flat, Microscale, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Replacement flat septa are designed for use with microscale reaction vials, or any other V-Bottom reaction vials.
These septa are constructed of white silicone rubber, bonded with a white PTFE facing. They will resist coring when punctured by a syringe needle. All septa are 3 mm thick.
Customers Who Bought This Also Bought