Reactor Heads with Ace-Thred Joints, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
Reactor heads feature a Duran style flange and Ace-Thred joints
They can be used with any reactor body with matching flange size. Compatible with pressure and filter reactors. Pressure rating is 35psi. Use a quick release clamp (not included) to connect to reactor body. Manufactured from borosilicate glass.
Ordering information: Available in three different flange sizes, with three different neck configurations. Clamp is not included.
Customers Who Bought This Also Bought