Stainless Steel Flat Top Worktable with Tubular Base, 14-Gauge Type 304, Eagle MHC™
Fournisseur: Eagle Group
Certificats
A propos de cet article
14 gauge type 304 stainless steel flat top.
- 35¹/₈ to 36¹/₈" adjustable working height
- 1⁵/₈" Ø legs and 1¼" Ø crossbracing are heavy gauge stainless steel
Rolled edge front and rear with ends turned under 90 degrees.
Ordering information: Accessories available include drawers and casters, sold separately.
Customers Who Bought This Also Bought