Stainless Steel Worktable with Backsplash and Undershelf, 16-Gauge Type 304, Eagle MHC™
Fournisseur: Eagle Group
Certificats
A propos de cet article
16 Gauge Type 304 stainless steel top with 4-¹/₂" backsplash at rear of table.
- Rolled edge front with ends turned under 90°
1-⁵/₈" diameter legs and adjustable undershelf are heavy gauge stainless steel. 35-¹/₈" to 36-¹/₈" adjustable working height.
.
Ordering information: Accessories available include drawers and casters, sold separately.
Customers Who Bought This Also Bought