Stainless Steel Cleanroom Tables, Solid Top, Eagle MHC™
Fournisseur: Eagle Group
Certificats
A propos de cet article
14-gauge, Type 304 stainless steel top.
- Stainless steel C- or H-base frame options
- Brushed stainless steel solid top
Ordering information: Must specify either a C- or H-frame at time of order. Tops are alcohol wiped and cleanroom bagged. Shipped knocked down reducing freight costs and potential for freight damage.
Customers Who Bought This Also Bought