VWR® Single-Lipped Atlas Shelves
Supplier: VWR International
Certificates
About this item
These standard shelves are powder-coated with high-quality, chemical-resistant epoxy powder and constructed of 16-gauge steel.
- Chemical-resistant epoxy powder coating
- Steel construction
- Single-lipped design
Designed to be infinitely adjustable along Atlas uprights, these lipped shelves can be used with the lip in the front or back and can be switched in the field. Shelf only.
Customers Who Bought This Also Bought