Gown Dispenser, Stainless Steel, Bandy
# de catalogue 89217-394
Fournisseur: Bandy
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Dispenser type:Wall-mountable
- No. of compartments:1
- PPE type:Garments
- Description:Gown dispenser
- Material:Stainless steel
- Supplier No.:DISP11002
Spécifications
A propos de cet article
Gown dispenser.
Hinged lid on top for easy refills. Open front for convenient visual gauge. Material is stainlesss steel 304. Electropolished.
Customers Who Bought This Also Bought