Garment Dispensers, Stainless Steel, Bandy
Fournisseur: Bandy
Certificats
A propos de cet article
Garment accessory dispenser for shoe covers, beard covers, bouffant caps/hats, ear protectors
Hinged lid on top for convenient refills. Slotted front for convenient visual gauge. Material is stainless steel 304. Electropolished.
Spécifications
- Catalog No:
- 89217-406
- 89217-398
- 89217-396
- 89217-400
- PPE type:
- Garments
- Garments
- Garments
- Garments
- Material:
- Stainless steel 304
- Stainless steel 304
- Stainless steel 304
- Stainless steel 304
- No. of compartments:
- 3
- 1
- 4
- 2
- Dispenser type:
- Wall-mountable
- Wall-mountable
- Wall-mountable
- Wall-mountable