Stainless Steel Flat Top Worktable with Undershelf, 16-Gauge Type 304, Eagle MHC™
Fournisseur: Eagle Group
Certificats
A propos de cet article
16 gauge type 304 stainless steel flat top.
- Adjustable undershelf are heavy gauge stainless steel
¹³/₈" diameter legs. Rolled edge front and rear with ends turned under 90°. 35.13 to 36.13" adjustable working height.
Ordering information: Accessories available include drawers and casters.
Customers Who Bought This Also Bought