Stainless Steel Flat Top Worktable with Undershelf, 14-Gauge Type 304, Eagle MHC™
Fournisseur: Eagle Group
Certificats
A propos de cet article
Tables have a 14 gauge type 304 stainless steel flat top.
- Non-adjustable
- Shipped knockdown
- For use with Eagle MHC worktables only
- Overshelves are available in single or double style
- Mounted on four stainless steel 1" Ø tubular base legs
1⁵/₈" Ø legs and adjustable undershelf are heavy gauge stainless steel. Rolled edge front and rear with ends turned under 90 degrees. 35¹/₈ to 36¹/₈" adjustable working height.
Ordering information: Available accessories include drawers and casters. It is recommended that overshelves be ordered when ordering tables. Specify choice of centered or rear-mounted on table. Otherwise table must be field drilled for mounting overshelves.
Customers Who Bought This Also Bought