Shaft Coupling, Stirring, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
The coupling connects the stirring motor shaft to the reactor stirring shaft.
The coupling has a flexible neoprene rubber body with a PTFE sleeve inside the corrosion-resistant metal end. This provides angular and parallel alignment of glass or metal stirring shafts without the danger of breakage. The rubber body absorbs shock and provides quiet, vibration-free operation with great torsional stiffness. Shaft coupling measures 4" overall.
Customers Who Bought This Also Bought