Mantle, Aluminum Housing, Bottom Opening, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Aluminum housed heating mantles feature a bottom opening to accommodate flasks with bottom valves
They can be used on benches with smaller sized flasks, or on floor or reactor stands with larger flasks. These mantles accommodate cylindrical and spherical flasks with bottom valves (see specifics below). Mantles must be operated through a temperature controller, and offer a temperature operating range of ambient + 10° to 450°C. Smaller sizes include a 122cm (4') detachable cord with interlocking connector. Larger sizes are supplied with a terminal box for hard wiring. CSA certified.
Customers Who Bought This Also Bought