Plate Support Shelf, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
Adjustable cast alloy, black epoxy coated support shelf
With the knob, the shelf measures 7x10". Fits up to 5/8" O.D. rod
Customers Who Bought This Also Bought